EMAIL THIS PAGE TO A FRIEND
Dermatitis : contact, atopic, occupational, drug
Contact allergy to a permanent marker.
PMID 22653131
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
60895 Gum rosin, natural resin