EMAIL THIS PAGE TO A FRIEND
TheScientificWorldJournal
Microbial transformations of 7-hydroxyflavanone.
PMID 22654578
Abstract
Microbial transformations of racemic 7-hydroxyflavanone using strains of genus Aspergillus (A. niger KB, A. niger 13/5, A. ochraceus 456) and the species Penicillium chermesinum 113 were studied. The products of O-methylation, O-methylation along with hydroxylation at C-3' and C-4', reduction of the carbonyl group, reduction of the carbonyl group along with hydroxylation at C-5, and dehydrogenation of C-2 and C-3 were obtained. Most of the products (with the exception of the O-methylation one) have stronger antioxidant properties than the initial substrate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
419753
7-Hydroxyflavanone, 98%
C15H12O3