EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
GH-independent IGF-I action is essential to prevent the development of nonalcoholic steatohepatitis in a GH-deficient rat model.
PMID 22659415