EMAIL THIS PAGE TO A FRIEND
Environmental science & technology
Tris(2,3-dibromopropyl) isocyanurate, hexabromocyclododecanes, and polybrominated diphenyl ethers in mollusks from Chinese Bohai Sea.
PMID 22663562
Abstract
A novel brominated flame retardant (BFR), tris(2,3-dibromopropyl) isocyanurate (TBC), as well as hexabromocyclododecanes (HBCDs) and polybrominated diphenyl ethers (PBDEs), were analyzed in 11 species of mollusks collected from nine coastal cities around the Chinese Bohai Sea in 2009 and 2010. The detection frequencies were 100%, 99%, and 77% for PBDEs, HBCDs, and TBC, respectively. Concentrations of ∑HBCDs ranged from below detection limit (nd) to 28.8 ng g(-1) on a dry weight (dw) basis, followed by ∑(12)PBDE (0.01-20.4 ng g(-1) dw) and TBC (nd-12.1 ng g(-1) dw). Statistically significant linear correlations were found among the three BFRs. Positive correlations were found between BFRs concentrations and lipid content in mollusks. The concentrations tend to decrease with increasing trophic levels of the mollusks, implying trophic dilution rather than biomagnifications of the BFRs in the aquatic food chains of the sampling area. Among the 11 mollusks species, Mytilus edulis showed higher bioaccumulation capability than others and was therefore considered to be an appropriate bioindicator of contamination by the BFRs in the Chinese Bohai Sea, in agreement with its previous selection for the biomonitoring of organochlorine pesticides (OCPs) and polychlorinated biphenyls (PCBs). A dramatic decrease in PBDE concentrations in mollusks of the area was found for the time period from 2003 to 2010, with a half-life of only 2.3 ± 1.7 years, reflecting a rapid response of mollusks to the change in pollution of the marine environment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
269999
Tris(2,3-dibromopropyl) isocyanurate, 97%
C12H15Br6N3O3