EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
A GIS-based generic real-time risk assessment framework and decision tools for chemical spills in the river basin.
PMID 22664261
Abstract
This paper presents a generic framework and decision tools of real-time risk assessment on Emergency Environmental Decision Support System for response to chemical spills in river basin. The generic "4-step-3-model" framework is able to delineate the warning area and the impact on vulnerable receptors considering four types of hazards referring to functional area, societal impact, and human health and ecology system. Decision tools including the stand-alone system and software components were implemented on GIS platform. A detailed case study on the Songhua River nitrobenzene spill illustrated the goodness of the framework and tool Spill first responders and decision makers of catchment management will benefit from the rich, visual and dynamic hazard information output from the software.