EMAIL THIS PAGE TO A FRIEND
Se pu = Chinese journal of chromatography
[Determination of target compounds in cefoperazone sodium and tazobactam sodium for injection by capillary electrophoresis].
PMID 22667102
Abstract
A capillary zone electrophoresis (CZE) method was developed for the simultaneous determination of cefoperazone sodium and tazobactam sodium in the injectable powder of cefoperazone sodium and tazobactam sodium with hydrochlorothiazide as the internal standard. The operation was carried out on a quartz capillary (75 cm x 75 microm i. d. , 63 cm effective length). The electrophoretic conditions were as follows: 40 mmol/L borax solution as the back ground electrolyte (BGE), 12. 0 kV applied voltage, 220 nm as the detection wavelength; the sample solution was injected by hydraulic pressure for 10 s at the height of 10 cm. The cefoperazone and tazobactam showed good linear relationship in the ranges of 0.25-3.96 g/L and 0.062-0.99 g/L with the correlation coefficients of 0.999 5 and 0.999 6, respectively. The relative standard deviations of relative peak areas were less than 3%. The preparation was stable in 208 min. The recovery results met the methodology requirements. The method is simple, rapid, reproducible, and suitable to control the quality of cefoperazone sodium and tazobactam sodium injectable powder.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4292
Cefoperazone sodium salt, 870-1015 μg/mg (anhydrous basis)
C25H26N9NaO8S2