EMAIL THIS PAGE TO A FRIEND
Sheng wu gong cheng xue bao = Chinese journal of biotechnology
[Expression, purification of proteasome subunit PSMB1 and application in screening of possible proteasome inhibitors].
PMID 22667125
Abstract
Proteasome is a multi-subunit protease complex in eukaryocytes, and plays an important role in ubiquitin-proteosome pathway. Recombinant proteasome can be used to screen proteasome inhibitors. In this study, recombinant plasmid of pET28a-PSMB1 was constructed by inserting human proteasome catalytic subunit (PSMB1) cDNA (726 bp) into the prokaryotic expression vector pET28a(+), and transforming the plasmid into E. coli BL21(DE3) cells for expression. After overnight induction (1 mmol/L IPTG, 20 degrees C), an expected protein band with molecular weight of 27 kDa was observed on SDS-PAGE gel. The recombinant protein was then purified through affinity chromatography, and the purity is more than 95%. The amino acid sequence of the recombinant protein was validated by NanoLC-MS/MS. The data from in vitro BIAcore analysis showed that the recombinant PSMB1 could bind to celastrol. The binding affinity between PSMB1 and 10 micromol/L celastrol was more than 27RU.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0869
Celastrol, ≥98% (HPLC), solid
C29H38O4