EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Highly controlled immortal polymerization of β-butyrolactone by a dinuclear indium catalyst.
PMID 22669203
Abstract
An ethoxy-bridged dinuclear indium catalyst was used for the ring opening polymerization of the cyclic ester β-butyrolactone to form the biodegradable polyester poly(hydroxybutyrate) (PHB). The catalyst shows remarkable activity and control during polymerization, allowing for formation of diblock polymers. Addition of high ratios of alcohols to the catalyst leads to fast chain transfer and immortal polymerization.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
219126
β-Butyrolactone, 98%
C4H6O2