EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Methods for the determination of plasma or tissue glutathione levels.
PMID 22669674
Abstract
We present two different methods for determining levels of glutathione in complex biological samples and plasma. The DTNB/GR enzyme recycling method is sensitive and requires no specialized equipment. The HPLC method is particularly useful for situations in which sample amounts are limited. Detailed instructions for performing each method as well as the advantages and disadvantages of each are discussed in this chapter.