EMAIL THIS PAGE TO A FRIEND
Indian journal of experimental biology
Regeneration of plantlets from mature embryo calli of Western Ghats land race cultivar of rice, Oryza sativa L.
PMID 22670480
Abstract
The Malnad region located in the Western Ghats of Karnataka is known for the cultivation of indigenous rain fed land race cultivar of rice. The present study was to investigate the callogenic and caulogenic potentialities of the two indigenous rice cultivar namely Karimundaga and Kanadatumba using dehusked mature embryo explants. For callus and shoot bud differentiation, the explants were cultured on Murashige and Skoog (MS) medium supplemented with 2,4-D (1-3 mg/L), IAA (1-2 mg/L), Kn (1-4 mg/L) and BAP (1-4 mg/L). The morphogenic potentialities of the two rice cultivar differed in texture of callus. In both the cultivar callogenic frequency was optimized at 1 mg/L 2,4-D concentration, it was 94% in Karimundaga and 58% in Kanadatumba. Supplementation of IAA either alone (1-2 mg/L) or in combination with Kn or BAP at 1 to 4 mg/L concentration of each induces shoot bud differentiation from the calli. In the cultivar Karimundaga caulogenic frequency was highest (10.60 +/- 2.55) at 1.0 mg/L IAA and 4.0 mg/L BAP concentration. While in the cultivar Kanadatumba highest number of shoot buds (7.90 +/- 2.69) was differentiated at 1.0 mg/L IAA and 4.0 mg/L Kn concentration. The calli derived regenerants were successfully acclimatized in the greenhouse and agro-morphological variations were evaluated. The growth characteristics and yield related parameters exhibited by in vitro plants were lower than the in vivo plants.