EMAIL THIS PAGE TO A FRIEND
Journal of fish diseases
Treatment of vibriosis in European sea bass larvae, Dicentrarchus labrax L., with oxolinic acid administered by bath or through medicated nauplii of Artemia franciscana (Kellogg): efficacy and residual kinetics.
PMID 22670590
Abstract
European sea bass larvae were challenged by bath with Listonella anguillarum strain 332A, 2.5×10(7) CFUmL(-1) for 1h. Fish either received no treatment or oral treatment with Artemia franciscana (Kellog) nauplii enriched with oxolinic acid, or bath treatments with oxolinic acid. Medication commenced 1day following challenge and was performed on days 1, 3 and 5 post-challenge at a dosage of 20mgL(-1) for 2h for bath treatments, while two doses each of 750 nauplii per fish were administered daily for five consecutive days in oral treatments. Cumulative mortality reached 96% for the unmedicated challenged group, 32% in the group receiving bath treatments and 17% in the group receiving medicated nauplii. Pharmacokinetic parameters of oxolinic acid were calculated in sea bass larvae, for both treatments. Steady-state concentrations of oxolinic acid of 48.0 and 75.2μgg(-1) were achieved for bath treatment and oral treatment, respectively, while the elimination half-life was calculated to be 25.1h for bath treatment and 21.7h for oral treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O0877
Oxolinic acid, quinolone antibiotic
C13H11NO5
67126
Oxolinic acid, analytical standard
C13H11NO5