EMAIL THIS PAGE TO A FRIEND
Organic letters
Improved protocol for indoline synthesis via palladium-catalyzed intramolecular C(sp2)-H amination.
PMID 22670815
Abstract
An efficient method has been developed for the synthesis of indoline compounds from picolinamide (PA)-protected β-arylethylamine substrates via palladium-catalyzed intramolecular amination of ortho-C(sp(2))-H bonds. These reactions feature high efficiency, low catalyst loadings, mild operating conditions, and the use of inexpensive reagents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I5605
Indoline, ReagentPlus®, 99%
C8H9N