EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Approach to the synthesis of indoline derivatives from diaryliodonium salts.
PMID 22670828
Abstract
An effective method of constructing the indoline moiety via intramolecular nucleophilic ring closure of a diaryliodonium salt is described. Diacetoxyiodoarene compounds (1a-1e) were converted into intermediate Koser's reagent and coupled with arylstannanes (7-10) to form diaryliodonium salts (11a-14e). Indoline compounds with different N-protecting groups, 15, 16, 17, and 18, were synthesized in higher yields by treating salts (11a-14e) with Cs(2)CO(3) and TEMPO. Regardless of the electronic environment of five para-substituted iodoarenes and the natures of four N-protected arylstannane groups, the conversion proceeded well to afford corresponding indolines in yields of 72-84 and 70-84%, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I5605
Indoline, ReagentPlus®, 99%
C8H9N