EMAIL THIS PAGE TO A FRIEND
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.]
Fractional carbon dioxide laser treatment to enhance skin permeation of ascorbic acid 2-glucoside with minimal skin disruption.
PMID 22672633
Abstract
Topical treatment with vitamin C has been used to treat photoaged skin and as a skin whitener, but no standard procedure exists for percutaneous delivery. To compare skin histology and the permeation of ascorbic acid 2-glucoside (AA2G) after fractional and conventional carbon dioxide (CO(2) ) laser pretreatment. The effect on porcine skin of treatment with different strengths of fractional and conventional CO(2) laser treatment was examined using scanning electron microscopy and transmission electron microscopy. Permeation of AA2G through porcine skin was tested in vitro using a Franz diffusion chamber. In vivo changes in fluorescein thiocyanate permeability in nude mice were examined using confocal laser scanning microscopy. Fractional CO(2) laser treatment with four or fewer passes caused less disruption than conventional laser treatment at the same fluence. AA2G permeation using four passes of fractional laser treatment was similar to that seen with conventional CO(2) laser treatment of the same fluence. Changes in permeability and in depth of permeation were higher with conventional than fractional laser treatment. Fractional CO(2) laser treatment can cause similar transdermal delivery of AA2G to conventional laser treatment with less skin disruption and a different pattern of histologic change.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1433
Ascorbyl Glucoside, Pharmaceutical Secondary Standard; Certified Reference Material
C12H18O11