EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Effects of chemical modification of sphingomyelin ammonium group on formation of liquid-ordered phase.
PMID 22672982
Abstract
Sphingomyelin (SM) and cholesterol form microdomains called lipid rafts in cellular membranes. To develop a versatile fluorescent lipid probe, chemical modifications to both the hydrophobic and hydrophilic portions of SM are essential. Few reports describing SM probes with a fluorophore at the polar head group have been published. This study examined the effect of substitution on an ammonium moiety of SM on the membrane properties of SM. Two SM analogs with small propargyl and allyl groups on the quaternary nitrogen atom were synthesized and subjected to analysis using differential scanning calorimetry, fluorescent anisotropy, detergent solubilization, surface pressure, and density measurements. Results demonstrated that the two SM analogs retained the membrane properties of SM, including formation of an ordered phase and the ability to interact with cholesterol. A dansyl-substituted SM was prepared for fluorescent measurements. Dansyl-SM showed less of a propensity to form microdomains. These findings imply the potential application of N-substituted SMs as a raft-specific molecular probe.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
39220
Dansyl chloride, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
C12H12ClNO2S