EMAIL THIS PAGE TO A FRIEND
Biomedical chromatography : BMC
Rapid screening and identification of α-glucosidase inhibitors from mulberry leaves using enzyme-immobilized magnetic beads coupled with HPLC/MS and NMR.
PMID 22674728
Abstract
α-Glucosidase plays important roles in the digestion and absorption of carbohydrates in the small intestine. The inhibition of α-glucosidase is regarded as a potential way to treat diabetes. We established an approach to screening α-glucosidase inhibitors from medicinal plants using enzyme-coated magnetic bead. Using 1-(3-dimethyl-aminopropyl)-3-ethylcarbodiimide and N-hydroxysuccinimide as reaction reagents, α-glucosidase was immobilized on the magnetic beads by covalent linkage. The conjugation of α-glucosidase to the magnetic beads was characterized using scanning electron microscope and X-ray diffractometer. The proposed approach was applied in fishing potential α-glucosidase inhibitors from extract of Morus alba, a Chinese medicinal plant. The structures of potential active compounds were identified via liquid chromatography-mass spectrometry and nuclear magnetic resonance. The results demonstrated that two flavonoids (isoquercitrin and astragalin) could bind to α-glucosidase, which was confirmed via conventional α-glucosidase inhibitory assay. Our findings suggested that enzyme-coated magnetic beads may be suitable for discovering active compounds from medicinal plants.