EMAIL THIS PAGE TO A FRIEND
Chirality
Enantioselective kinetics of α-hexachlorocyclohexane in earthworm (Eisenia fedtia) and forest soil.
PMID 22674802
Abstract
The enantioselective bioaccumulation and elimination behaviors of α-hexachlorocyclohexane (α-HCH) enantiomers in earthworm and soil were investigated by chiral gas chromatography. Enantiomer fraction values were calculated as indicators of the enantioselectivity. The mature earthworms were exposed to 0.10 µg g(-1)(wwt) (0.14 µg g(-1)(dwt)) spiked soil continuously for the bioaccumulation, and the elimination was conducted after an enrichment period in the soil. The results showed that both the bioaccumulation and elimination processes followed monophasic kinetics, body residues of α-HCH in earthworm increased to high level at the fifth day, and enantioselectivity was found in the bioaccumulation process with the rate constant (k) of 0.80 d(-1) for (+)-α-HCH and 0.74 d(-1) for (-)-α-HCH. The half life (t(1/2)) of the enantiomers obtained in the elimination process was within one day. The bioaccumulation factors of steady state of α-HCH enantiomers were 2.82 for (+)-α-HCH and 2.75 for (-)-α-HCH. The enantiomer fractions of earthworm and soil obviously below 0.5 during uptake and elimination processes indicate significant enantioselectivity and preferential depuration of (+)-α-HCH in earthworm. However, earthworms do not have a great capacity for getting rid of α-HCH in polluted soil shown by a contradistinctive experiment.