EMAIL THIS PAGE TO A FRIEND
The Israel Medical Association journal : IMAJ
Drug-induced palate osteonecrosis following nasal surgery.
PMID 22675864
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2371
Pamidronate disodium salt hydrate, ≥95% (NMR), solid
C3H9NO7P2Na2 · xH2O