EMAIL THIS PAGE TO A FRIEND
International journal of biological macromolecules
Structural analysis of Herbaspirillum seropedicae lipid-A and of two mutants defective to colonize maize roots.
PMID 22676993
Abstract
Lipid-A was isolated by mild acid hydrolysis from lipopolysaccharides extracted from cells of Herbaspirillum seropedicae, strain SMR1, and from two mutants deficient in the biosynthesis of rhamnose (rmlB⁻ and rmlC⁻). Structural analyzes were carried out using MALDI-TOF and derivatization by per-O-trimethylsilylation followed by GC-MS in order to determine monosaccharide and fatty acid composition. De-O-acylation was also performed to determine the presence of N-linked fatty acids. Lipid-A from H. seropedicae SMR1 showed a major structure comprising 2-amino-2-deoxy-glucopyranose-(1→6)-2-amino-2-deoxy-glucopyranose phosphorylated at C4' and C1 positions, each carrying a unit of 4-amino-4-deoxy-arabinose. C2 and C2' positions were substituted by amide-linked 3-hydroxy-dodecanoic acids. Both rhamnose-defective mutants showed similar structure for their lipid-A moieties, except for the lack of 4-amino-4-deoxy-arabinose units attached to phosphoryl groups.