EMAIL THIS PAGE TO A FRIEND
FEBS letters
Alendronate promotes plasmin-mediated MMP-9 inactivation by exposing cryptic plasmin degradation sites within the MMP-9 catalytic domain.
PMID 22677171
Abstract
Irreversible MMP-9 inhibition is considered a significant therapeutic goal in inflammatory, vascular and tumour pathology. We report that divalent cation chelators Alendronate and EDTA not only directly inhibited MMP-9 but also promoted irreversible plasmin-mediated MMP-9 inactivation by exposing cryptic plasmin-degradation sites within the MMP-9 catalytic-domain and producing an inhibitory hemopexin-domain fragment. This effect was also observed using MDA-MB-231 breast cancer cells, which activated exogenous plasminogen to degrade endogenous proMMP-9 in the presence of Alendronate or EDTA. Degradation-mediated inactivation of proMMP-9 occurred in the absence of transient activation, attesting to the incapacity of plasmin to directly activate proMMP-9 and direct MMP-9 inhibition by Alendronate and EDTA. Our study provides a novel rational for therapeutic Alendronate use in MMP-9-dependent pathology characterised by plasminogen activation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H9291 Hemopexin from human plasma, ≥95% (SDS-PAGE), lyophilized powder