EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Development, validation and comparison of two microextraction techniques for the rapid and sensitive determination of pregabalin in urine and pharmaceutical formulations after ethyl chloroformate derivatization followed by gas chromatography-mass spectrometric analysis.
PMID 22677651
Abstract
The present article reports first time the use of solid-phase microextraction (SPME) and dispersive liquid-liquid microextraction (DLLME) to extract pregabalin (PRG) from urine and pharmaceutical formulations followed by GC-MS analysis after ethyl chloroformate (ECF) derivatization. PRG is an antiepileptic and analgesic drug, which is a structural analogue of γ-amino-butyric acid (GABA). It is approved by Food and Drug Administration (FDA) for the treatment of central nervous system (CNS) disorders and neuropathic pain. Initially PRG was derivatized with ECF in the presence of pyridine at room temperature for 30s. Experimental parameters were investigated for derivatization, SPME and DLLME conditions. The limit of detection (LOD) and limit of quantitation (LOQ) were found to be 0.019 μg/ml and 0.063 μg/ml for SPME and 0.022 μg/ml and 0.075 μg/ml for DLLME respectively. The percentage recovery, in case of SPME was in the range of 83-98% while for DLLME it is in the range of 84-98%. The intra and inter-day precisions were found to be less than 6%. The developed methods after ECF derivatization were found to be simple, fast, efficient and inexpensive. DLLME has several advantages like lesser extraction time and cost effectiveness as compared to SPME. The developed methods may find wide application for the routine determination of PRG in biological as well as in quality control samples of pharmaceutical formulations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
185892
Ethyl chloroformate, 97%
C3H5ClO2
57317 SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 60 μm (partially crosslinked phase), for use with autosampler/HPLC, needle size 24 ga, StableFlex fiber
57310-U SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm (partially crosslinked phase), needle size 24 ga, for use with manual holder
57346-U SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), fused silica fiber, df 65 μm (partially crosslinked phase), for use with manual holder, needle size 23 ga
57311 SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm (partially crosslinked phase), needle size 24 ga, for use with autosampler
57345-U SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm (partially crosslinked phase), for use with autosampler, needle size 23 ga
57326-U SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm, needle size 24 ga, StableFlex, for use with manual holder
57327-U SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm, needle size 24 ga, StableFlex, for use with autosampler
57293-U SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm, needle size 23 ga, PDMS/DVB StableFlex, for use with autosampler
57902-U SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm (bonded phase), for use with autosampler, needle size 23 ga, metal alloy fiber
FFA57293-U SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm, needle size 23 ga, Stableflex, for use with multi fiber exchanger
FFA57327-U SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm, for use with multi fiber exchanger, needle size 24 ga, Stableflex
57359-U SPME Portable Field Sampler, coating Polydimethylsiloxane/Divinylbenzene (PDMS/DVB)