EMAIL THIS PAGE TO A FRIEND
Biotechnology journal
Molecular dynamics simulation of the induced-fit binding process of DNA aptamer and L-argininamide.
PMID 22678933
Abstract
Aptamers are rare functional nucleic acids with binding affinity to and specificity for target ligands. Recent experiments have lead to the proposal of an induced-fit binding mechanism for L-argininamide (Arm) and its binding aptamer. However, at the molecular level, this mechanism between the aptamer and its coupled ligand is still poorly understood. The present study used explicit solvent molecular dynamics (MD) simulations to examine the critical bases involved in aptamer-Arm binding and the induced-fit binding process at atomic resolution. The simulation results revealed that the Watson-Crick pair (G10-C16), C9, A12, and C17 bases play important roles in aptamer-Arm binding, and that binding of Arm results in an aptamer conformation optimized through a general induced-fit process. In an aqueous solution, the mechanism has the following characteristic stages: (a) adsorption stage, the Arm anchors to the binding site of aptamer with strong electrostatic interaction; (b) binding stage, the Arm fits into the binding site of aptamer by hydrogen-bond formation; and (c) complex stabilization stage, the hydrogen bonding and electrostatic interactions cooperatively stabilize the complex structure. This study provides dynamics information on the aptamer-ligand induced-fit binding mechanism. The critical bases in aptamer-ligand binding may provide a guideline in aptamer design for molecular recognition engineering.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3913
L-Argininamide dihydrochloride
C6H15N5O · 2HCl