EMAIL THIS PAGE TO A FRIEND
Ukrains'kyi biokhimichnyi zhurnal (1999 )
[Antioxidant properties of proteins after freezing-thawing].
PMID 22679758
Abstract
Experimental data are presented which were obtained under comparative evaluation of influence of different freezing-thawing conditions on antioxidant properties of isolated proteins: human serum albumin, cytochrome c from the horse heart and glucose oxidase from Aspergillus niger. The observed protein antioxidant activity alterations are assumed to be a result of protein conformational changes. The character of freezing-thawing influence on the protein antioxidant activity depends on the molecular structure and cooling conditions.