EMAIL THIS PAGE TO A FRIEND
Renal failure
Hair dye related acute kidney injury--a clinical and experimental study.
PMID 22680304
Abstract
We studied paraphenylenediamine (PPD)-related acute kidney injury (AKI) in 81 patients and also in albino rats experimentally. In the patients' group AKI was found in 32.7%. Of them, 81.4% needed dialysis support. The overall mortality was 25.9%. In experimental rats the renal lesions were noted in all and they were glomerular congestion, intertubular (interstitial) hemorrhages, acute tubular necrosis, mesangial proliferation, and intratubular casts. The severity of renal injury appears to be dose dependent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
78460
1,4-Phenylenediamine dihydrochloride, ≥99.0%
C6H8N2 · 2HCl