EMAIL THIS PAGE TO A FRIEND
Xenobiotica; the fate of foreign compounds in biological systems
Isolation and identification of the metabolites of paeonol in human urine.
PMID 22680310
Abstract
Paeonol, a major component of Paeonia suffruticosa Andrews, is used in clinical situations in China as a natural anti-inflammatory agent. The aim of the present study is to investigate the metabolism of paeonol in humans. Six metabolites were isolated from human urine after oral administration of paeonol, and their structures were elucidated as resacetophenone (M1), resacetophenone-2-O-sulfate (M2), 2-hydroxy-4-methoxyacetophenone-5-O-sulfate(M3), 2-hydroxy-4-methoxyacetophenone-5-O-glucopyranuronoside (M4), 2-hydroxyacetophenone-4-O-glucopyranuronoside (M5) and 2,5-dihydroxy-4-methoxyacetophenone(M6) by a series of analyses involving mass spectrometry, (1)H NMR, (13)C NMR and NOESY spectra. In addition, three more metabolites 2,4-dihydroxyacetophenone-5-O-sulfate (M7), paeonol-2-O-glucopyranuronoside (M8) and paeonol-2-O-sulfate (M9), were identified in human urine by using a UPLC/Q-TOF-MS/MS method. This is the first study of paeonol metabolism in humans. Based on the identified metabolites, possible metabolic pathways of paeonol in humans are proposed. Paeonol is metabolized mainly by hydroxylation and demethylation to give the corresponding phase I metabolites, M1, M6 and 2,4,5-trihydroxyacetophenone, and which then underwent conjugation with glucuronic acid or sulfuric acid to form phase II metabolites.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H35803
2′-Hydroxy-4′-methoxyacetophenone, 99%
C9H10O3