EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
IDH1 and IDH2 have critical roles in 2-hydroxyglutarate production in D-2-hydroxyglutarate dehydrogenase depleted cells.
PMID 22683334
Abstract
D-2-hydroxyglutaric aciduria (D-2HGA) is a hereditary metabolic disorder characterized by the elevated levels of D-2-hydroxyglutaric acid (D-2HG) in urine, plasma and cerebrospinal fluid. About half of the patients have autosomal recessive mutations in D-2-hydroxyglutarate dehydrogenase (D2HGDH) gene. To analyze the origin of D-2HG in D2HGDH-depleted cells, we used small interfering RNA (siRNA) techniques. We found that knockdown of D2HGDH in MCF7 cells increased the levels of 2HG, mimicking D2HGDH mutant cells. Additional knockdown of isocitrate dehydrogenase 1 (IDH1) or isocitrate dehydrogenase 2 (IDH2) decreased the level of 2HG in D2HGDH knockdown MCF7 cells. Conversely, ectopic expression of IDH1 or IDH2 increased 2HG in MCF7 cells. These results suggest that IDH1 and IDH2 have roles in production of D-2HG in cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
94577
DL-α-Hydroxyglutaric acid disodium salt, ≥95.0% (GC)
C5H6Na2O5