EMAIL THIS PAGE TO A FRIEND
Progress in neuro-psychopharmacology & biological psychiatry
The recreational tryptamine 5-MeO-DALT (N,N-diallyl-5-methoxytryptamine): a brief review.
PMID 22683457
Abstract
5-MeO-DALT (N,N-diallyl-5-methoxytryptamine) is a psychoactive substance, sold primarily over the Internet as a 'research chemical' or 'plant food'. Although details for the synthesis of this tryptamine have been available since 2004, its use as a hallucinogenic drug has been reported only occasionally in on-line user fora. It is controlled in only a few countries world-wide. There is little scientifically-based literature on the pharmacological, physiological, psychopharmacological, toxicological and epidemiological characteristics of 5-MeO-DALT. Here we review what is known about these aspects. We also report what we believe to be the first death involving the use of this substance. The case involved a man in his mid-20s who died in mid-2010. The coroner concluded that the deceased "died from injuries sustained after being hit by a lorry whilst under the influence of 5-MeODALT". It is critical that any other cases, including non-fatal instances, are documented so that a scientific evidence-base can be established for this drug.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
286583
5-Methoxytryptamine, 97%
C11H14N2O