EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Molecular recognitive adsorption of aqueous tetramethylammonium on the organic derivative of Hiroshima University Silicate-1 with a silane coupling reagent.
PMID 22684207
Abstract
Hiroshima University Silicate-1 (HUS-1), composed of silicate sheets with a halved sodalite cage and the interlayer tetramethylammonium (TMA) cation in the cage, was modified with dimethyldichlorosilane to form the organic derivative in which a dimethyl group was grafted onto the interlayer surface and a part of the TMA was removed, and the silylated HUS-1 effectively and selectively adsorbed TMA from water even in the presence of aqueous phenol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
87708
Tetramethylammonium bromide, for ion pair chromatography, ≥99.0% (AT)
C4H12BrN
426296
Tetramethylammonium bromide, ACS reagent, ≥98.0%
C4H12BrN
195758
Tetramethylammonium bromide, 98%
C4H12BrN
87709
Tetramethylammonium bromide, for electrochemical analysis, ≥99.0%
C4H12BrN
87718
Tetramethylammonium chloride, BioUltra, for molecular biology, ≥99.0% (AT)
C4H12ClN
74202
Tetramethylammonium chloride, for ion pair chromatography, ≥99.0% (AT)
C4H12ClN
T19526
Tetramethylammonium chloride, reagent grade, ≥98%
C4H12ClN
T3411
Tetramethylammonium chloride solution, for molecular biology
459135
Tetramethylammonium fluoride, 97%
C4H12FN
T7505
Tetramethylammonium hydroxide pentahydrate, ≥97%
C4H13NO · 5H2O
87741
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
C4H13NO · 5H2O
426318
Tetramethylammonium hydroxide solution, ACS reagent
C4H13NO
334901
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
C4H13NO
331635
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
C4H13NO
328251
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
C4H13NO
235946
Tetramethylammonium iodide, 99%
C4H12IN
425257
Tetramethylammonium nitrate, 95%
C4H12N2O3
02799
Tetramethylammonium sulfate, for ion pair chromatography, ≥99.0% (T)
C8H24N2O4S
87743
Tetramethylammonium sulfate hydrate
C8H24N2O4S · xH2O