EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Calciate-mediated intermolecular hydroamination of diphenylbutadiyne with secondary anilines.
PMID 22684250
Abstract
Calciate-mediated intermolecular hydroamination of diphenylbutadiyne with N-phenyl and N-isopropyl-substituted anilines yields E- and Z-isomers of the corresponding 1-anilino-1,4-diphenylbut-1-ene-3-yne. In the case of HNPh(2) solely heterobimetallic K(2)Ca(NPh(2))(4) is able to effectively catalyze this hydroamination reaction in tetrahydrofuran at elevated temperatures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
161233
1,4-Diphenylbutadiyne, 99%
C16H10