EMAIL THIS PAGE TO A FRIEND
Journal of toxicology and environmental health. Part A
Development of a strategy for biological monitoring in a chemical plant producing 3,3'-dichlorobenzidine dihydrochloride.
PMID 22686315
Abstract
In a chemical plant in Germany producing 3,3'-dichlorobenzidine dihydrochloride for the manufacture of colorants, blood and urine samples were taken for biological monitoring. 3,3'-Dichlorobenzidine (DBZ) was analyzed in urine by thin-layer chromatography and subsequently further combined with analysis of adducts of 3,3'-DBZ in hemoglobin. Data highlight current ranges of industrial exposure to 3,3'-DBZ in Germany and demonstrate the applicability of biological monitoring to minimize this exposure. Effective biological monitoring was achieved by a combination of monitoring hemoglobin adducts with spot samplings of urinary 3,3'-DBZ excretion in cases of reported exposure periods. Data presented might help to identify biological guidance values (BGV/BAR) for 3,3'-DBZ-exposed individuals.