EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Use of rice straw biochar simultaneously as the sustained release carrier of herbicides and soil amendment for their reduced leaching.
PMID 22686323
Abstract
The sustained release and reduced leaching of herbicides is expected for enhancing their efficacy and minimizing their pollution. For this purpose, the rice straw biochar made at a relatively low temperature (350 °C) (RS350) was used simultaneously as the carrier for incorporating herbicides besides as the soil amendment. In this way, the sustained release of herbicides acetochlor and 2,4-D was obtained in the release experiments, due to the high and reversible sorption by RS350 biochar. Besides, the RS350 biochar significantly reduced the leached amount of herbicides by 25.4%-40.7% for acetochlor, and by 30.2%-45.5% for 2,4-D, depending on the depth (50 or 100 mm) of biochar-amended soil horizon. The high retention of both herbicides in the biochar-amended topsoil makes it possible to extend their efficacy. The results suggest a potential way of using low temperature biochars to reduce the leaching of herbicides without impacting their efficacy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33379
Acetochlor, PESTANAL®, analytical standard
C14H20ClNO2