EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Ring expansion of cyclic β-amino alcohols induced by diethylaminosulfur trifluoride: synthesis of cyclic amines with a tertiary fluorine at C3.
PMID 22686336
Abstract
As the replacement of a hydrogen atom by a fluorine atom in a compound can have an important impact on its biological properties, the development of methods allowing the introduction of a fluorine atom is of great importance. The scope and limitations of the ring expansion of cyclic 2-hydroxymethyl amines induced by diethylaminosulfur trifluoride (DAST) to produce cyclic β-fluoro amines was studied as well as the enantioselectivity of the process.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
235253
(Diethylamino)sulfur trifluoride, 95%
C4H10F3NS