EMAIL THIS PAGE TO A FRIEND
Biological & pharmaceutical bulletin
β-Amyrin oxidation by oat CYP51H10 expressed heterologously in yeast cells: the first example of CYP51-dependent metabolism other than the 14-demethylation of sterol precursors.
PMID 22687421
Abstract
CYP51 has been recognized as a unique CYP family that consists of one isolated molecular species, a sterol 14-demethylase essential for sterol biosynthesis. However, another CYP51 gene classified as the CYP51H subfamily has been identified in higher plants, in addition to a sterol 14-demethylase gene, CYP51G1. To shed light on the function of this "second CYP51", oat CYP51H10 was introduced into the β-amyrin-producing yeast cells, and the effect of the expressed CYP51H10 on β-amyrin metabolism in the host cells was examined. In the CYP51H10-introduced cells, β-amyrin was converted to a metabolite with 12,13-epoxy and one additional hydroxyl group. Since the 12,13-epoxy group introduced into β-amyrin ring is an essential structure of avenacin A-1, a triterpene glycoside produced in oat from β-amyrin, the present findings indicate the contribution of CYP51H10 to avenacin A-1 biosynthesis from β-amyrin. This is the first study showing a second function of the CYP51 family.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
53017
α-Amyrin, analytical standard
C30H50O
09236
β-Amyrin, analytical standard
C30H50O