EMAIL THIS PAGE TO A FRIEND
Internal medicine (Tokyo, Japan)
A case of diaphragmatic paralysis complicated by herpes-zoster virus infection.
PMID 22687801
Abstract
Diaphragmatic paralysis is commonly caused by surgical and traumatic injuries, malignant neoplasm, and neurodegenerative disorders. However, in rare instances, diaphragmatic paralysis due to herpes-zoster virus infection has been reported. Here, we describe an 85-year-old woman who developed left hemidiaphragmatic paralysis within 19 days of the appearance of a typical herpes-zoster rash involving the C4-5 dermatome on the left side. Clinical and radiological findings revealed no local causes of phrenic nerve lesion. The hemidiaphragmatic paralysis was thought to be caused by herpes-zoster virus infection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F7932
Famciclovir, ≥98% (HPLC)
C14H19N5O4