EMAIL THIS PAGE TO A FRIEND
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan
[Performance evaluation of a fluorescamine-HPLC method for determination of histamine in fish and fish products].
PMID 22688027
Abstract
A method for the quantification of histamine in fish and fish products using tandem solid-phase extraction and fluorescence derivatization with fluorescamine was previously developed. In this study, we improved this analytical method to develop an official test method for quantification of histamine in fish and fish products, and performed a single laboratory study to validate it. Recovery tests of histamine from fillet (Thunnus obesus), and two fish products (fish sauce and salted and dried whole big-eye sardine) that were spiked at the level of 25 and 50 µg/g for T. obesus, and 50 and 100 µg/g for the two fish products, were carried out. The recoveries of histamine from the three samples tested were 88.8-99.6% with good repeatability (1.3-2.1%) and reproducibility (2.1-4.7%). Therefore, this method is acceptable for the quantification of histamine in fish and fish products. Moreover, surveillance of histamine content in food on the market was conducted using this method, and high levels of histamine were detected in some fish products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F9015
Fluorescamine, ≥98% (TLC), powder, used for detection of primary amines
C17H10O4