EMAIL THIS PAGE TO A FRIEND
Inflammation
Celecoxib enhances the anti-inflammatory effects of farnesylthiosalicylic acid on T cells independent of prostaglandin E(2) production.
PMID 22688643
Abstract
Celecoxib (Celebrex(®)), a non-steroidal anti-inflammatory drug and selective cyclooxygenase-2 inhibitor, is widely used to treat arthritis and other inflammatory disorders. Awareness of its anti-proliferative properties has prompted another indication for its use, in preventing colon polyps in high-risk populations. Farnesylthiosalicylic acid (FTS; Salirasib(®)), designed to inhibit oncogenic Ras and currently under evaluation in phase I/II and II clinical trials, was recently shown by our group to exert anti-inflammatory effects on both lymphocytes and mast cells. Here we examined whether celecoxib combined with FTS would enhance this anti-inflammatory activity. While each drug separately inhibited Ras activation in these cells, their combination yielded more marked inhibition as well as further inhibition of ERK phosphorylation, lymphocyte adhesion, and interleukin-2 secretion. The inhibitory effects, moreover, were independent of prostaglandin E(2) secretion. These data point to the promising potential of combined treatment with celecoxib and FTS for inflammatory disorders involving lymphocytes.