EMAIL THIS PAGE TO A FRIEND
Nature reviews. Nephrology
Childhood nephrotic syndrome--current and future therapies.
PMID 22688744
Abstract
The introduction of corticosteroids more than 50 years ago dramatically improved the prognosis of children with nephrotic syndrome. Corticosteroids remain the standard initial treatment for children with this disease, but a considerable proportion of patients do not respond and are therefore at risk of progressing to end-stage renal disease. Because of this risk, new therapeutic strategies are needed for steroid-resistant nephrotic syndrome. These strategies have historically focused on identifying effective alternative immunosuppressive agents, such as ciclosporin and tacrolimus, yet evidence now indicates that nephrotic syndrome results from podocyte dysfunction. Even conventional immunosuppressive agents, such as glucocorticoids and ciclosporin, directly affect podocyte structure and function, challenging the 'immune theory' of the pathogenesis of childhood nephrotic syndrome in which disease is caused by T cells. This Review summarizes the currently available treatments for childhood nephrotic syndrome, and discusses selected novel pathways in podocytes that could be targeted for the development of next-generation treatments for children with this syndrome.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M3047
Mizoribine, ≥98% (TLC)
C9H13N3O6