EMAIL THIS PAGE TO A FRIEND
Journal of mass spectrometry : JMS
GC-MS(n) and LC-MS/MS couplings for the identification of degradation products resulting from the ozonation treatment of Acetochlor.
PMID 22689619
Abstract
The degradation of the chloracetamide herbicide acetochlor has been studied under simulated ozonation treatment plant conditions. The degradation of acetochlor included the formation of several degradation products that were identified using GC/ion-trap mass spectrometry with EI and CI and HPLC/electrospray-QqTOF mass spectrometry. Thirteen ozonation products of acetochlor have been identified. Ozonation of the deuterated herbicide combined to MS(n) and high-resolution mass measurement allowed effective characterization of the degradation products. At the exception of one of them, the product B (2-chloro-2', ethyl-6', methyl-acetanilide), none of the identified degradation products has been already reported in the literature. Post-ozonation kinetics studies revealed that the concentrations of most degradation products evolved noticeably with time, particularly during the first hours following the ozonation treatment. This raises concerns about the fate of degradation products in the effluents of treatment plants and suggests the need for a better control on these products if their toxicity was demonstrated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33379
Acetochlor, PESTANAL®, analytical standard
C14H20ClNO2