EMAIL THIS PAGE TO A FRIEND
Journal of clinical oncology : official journal of the American Society of Clinical Oncology
Late and prolonged pseudoprogression in glioblastoma after treatment with lomustine and temozolomide.
PMID 22689800
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L5918
Lomustine, ≥98%
C9H16ClN3O2