EMAIL THIS PAGE TO A FRIEND
Journal of chromatographic science
Validation of an ultra-high performance liquid chromatography tandem mass spectrometric method for quantifying uracil and 5,6-dihydrouracil in human plasma.
PMID 22689904
Abstract
A simple, rapid, sensitive and specific ultra-high-performance liquid chromatography-tandem mass spectrometry method (Waters UPLC-MS-MS) is developed and validated for the quantification of uracil (U) and 5,6-dihydrouracil (UH2) levels in human plasma. Analytes are extracted using ethyl acetate and isopropanol after deproteination, and separated by high-performance liquid chromatography (HPLC) (Acquity UPLC BEH C18 column) in a binary mobile phase system under gradient elution conditions at a flow rate of 0.4 mL/min. 5-Bromo-uracil (UBr) is used as the internal standard. The detection is performed on a triple-quadrupole mass spectrometer via electrospray positive ionization. Multiple reaction monitoring mode using the transitions m/z 112.82 → 70.05, m/z 114.88 → 55.04 and m/z 190.83 → 117.86 is used to quantify U, UH2 and UBr, respectively. The method is linear in the concentration range of 0.625-160.0 ng/mL. The total run time is 4.5 min per injection. Nine-point calibration curve and four-points quality controls are used. Excellent linearity and precision are observed with correlation coefficient (r(2)) > 0.9999. The intra-batch and inter-batch precisions are ≤ 7.3% and ≤ 8.6%, and accuracy is ≤ 17%. The developed method is shown to be suitable for routine quantitative determination of U, UH2 and 5,6-dihydrouracil-to-uracil ratio in clinical practice.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7628
Dihydrouracil
C4H6N2O2