EMAIL THIS PAGE TO A FRIEND
Circulation
When the heart runs out of heartbeats: treatment options for refractory end-stage heart failure.
PMID 22689932
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M4659
Milrinone, ≥97% (TLC), powder
C12H9N3O