EMAIL THIS PAGE TO A FRIEND
Journal of biomedicine & biotechnology
Curcumin attenuates gastric cancer induced by N-methyl-N-nitrosourea and saturated sodium chloride in rats.
PMID 22690125
Abstract
To determine effects of curcumin on N-methyl-N-nitrosourea (MNU) and saturated sodium chloride (s-NaCl)-induced gastric cancer in rats. Male Wistar rats were divided into 5 groups: control (CO), control supplemented with 200 mg/kg curcumin (CC), MNU + s-NaCl, MNU + s-NaCl supplemented with 200 mg/kg curcumin daily for the first 3 weeks (MNU + s-NaCl + C3W), and MNU + s-NaCl supplemented with curcumin for 20 weeks (MNU + s-NaCl + C20W). To induce stomach cancer, rats except for CO and CC were orally treated with 100 mg/kg MNU on day 0 and 14, and s-NaCl twice-a-week for the first 3 weeks. The experiment was finished and rats were sacrificed at the end of 20 weeks. Cancers were found in forestomachs of all rats in MNU + s-NaCl. The expressions of phosphorylated inhibitor kappaB alpha (phospho-IκBα), 8-hydroxy-2'-deoxyguanosine (8-OHdG), and cyclin D1 significantly increased in MNU + s-NaCl compared with CO. Curcumin treatments for 3 and 20 weeks reduced the cancer incidence resulting in a decrease of phospho-IκBα expression in benign tumor-bearing rats compared with MNU + s-NaCl. Curcumin treatment for 20 weeks also decreased 8-OHdG expression in benign tumor-bearing rats compared with MNU + s-NaCl. Curcumin can attenuate cancer via a reduction of phospho-IκBα and 8-OHdG expressions, which may play a promising role in gastric carcinogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1517
N-Nitroso-N-methylurea, ISOPAC®
C2H5N3O2
N4766
N-Nitroso-N-methylurea, Bulk package
C2H5N3O2