EMAIL THIS PAGE TO A FRIEND
Scandinavian journal of immunology
The cystine/glutamate antiporter regulates the functional expression of indoleamine 2,3-dioxygenase in human dendritic cells.
PMID 22690871
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
219746
DL-Homocysteic acid, 97%
C4H9NO5S
H9633
L-Homocysteic acid
C4H9NO5S