EMAIL THIS PAGE TO A FRIEND
Annual review of chemical and biomolecular engineering
Chemical processing of materials on silicon: more functionality, smaller features, and larger wafers.
PMID 22691090
Abstract
The invention of the transistor followed by more than 60 years of aggressive device scaling and process integration has enabled the global information web and subsequently transformed how people communicate and interact. The principles and practices built upon chemical processing of materials on silicon have been widely adapted and applied to other equally important areas, such as microfluidic systems for chemical and biological analysis and microscale energy storage solutions. The challenge of continuing these technological advances hinges on further improving the performance of individual devices and their interconnectivity while making the manufacturing processes economical, which is dictated by the materials' innate functionality and how they are chemically processed. In this review, we highlight challenges in scaling up the silicon wafers and scaling down the individual devices as well as focus on needs and challenges in the synthesis and integration of multifunctional materials.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF37993595
Silicon, rod, 100mm, diameter 10.0mm, single crystal - random orientation, 100%
Si
GF64686998
Silicon, rod, 100mm, diameter 2.0mm, crystalline, 100%
Si
GF22929307
Silicon, rod, 100mm, diameter 5.0mm, crystalline, 100%
Si
GF16363391
Silicon, rod, 10mm, diameter 2.0mm, crystalline, 100%
Si
GF66465276
Silicon, rod, 25mm, diameter 2.0mm, crystalline, 100%
Si
GF43555664
Silicon, rod, 25mm, diameter 3.15mm, single crystal - random orientation, 100%
Si
GF70438518
Silicon, rod, 40mm, diameter 20mm, single crystal, -100, 99.999%
Si
GF35274113
Silicon, rod, 40mm, diameter 20mm, single crystal, -111, 99.999%
Si
GF05482852
Silicon, rod, 50mm, diameter 2.0mm, crystalline, 100%
Si
GF91515034
Silicon, rod, 50mm, diameter 3.15mm, single crystal - random orientation, 100%
Si
GF14076930
Silicon, rod, 50mm, diameter 5.0mm, crystalline, 100%
Si
GF28448691
Silicon, rod, 50mm, diameter 5mm, single crystal, -100, 99.999%
Si
GF40945754
Silicon, rod, 50mm, diameter 5mm, single crystal, -111, 99.999%
Si
GF64342188
Silicon, rod, 50mm, diameter 6mm, single crystal, -111, 99.999%
Si
GF10907377
Silicon, rod, 80mm, diameter 20mm, single crystal, -100, 99.999%
Si
GF91141792
Silicon, sheet, 10x10mm, thickness 0.5mm, single crystal, -100, 100%
Si
GF71817543
Silicon, sheet, 10x10mm, thickness 0.6mm, single crystal, -100, 100%
Si
GF75683212
Silicon, sheet, 14x14mm, thickness 1.0mm, single crystal, -100, 100%
Si
GF84840004
Silicon, sheet, 14x14mm, thickness 1.0mm, single crystal, -111, 100%
Si
GF20311254
Silicon, sheet, 25x25mm, thickness 1.0mm, polycrystalline, 99.999%
Si
GF41762493
Silicon, sheet, 25x25mm, thickness 1.0mm, single crystal, -100, 100%
Si
GF85621923
Silicon, sheet, 40x40mm, thickness 3.0mm, single crystal, p-type, 100%
Si
GF73529051
Silicon, sheet, 52x52mm, thickness 1.0mm, polycrystalline, 99.999%
Si
GF23955622
Silicon, single crystal, -100, n-type, phosphorus doped, diameter 150mm, length 0.65mm, 100%
Si
GF06693756
Silicon, single crystal, -111, n-type, phosphorus doped, diameter 76.2mm, length 0.50mm, 100%
Si
GF68832805
Silicon, single crystal, -111, n-type, phosphorus doped, diameter 76.2mm, length 0.75mm, 100%
Si
GF52781520
Silicon, sphere, 10pcs, diameter 2.0mm, precision sphere grade 25
Si
GF68982834
Silicon, sphere, 20pcs, diameter 2.0mm, precision sphere grade 25
Si
GF54682050
Silicon, sphere, 50pcs, diameter 2.0mm, precision sphere grade 25
Si
GF72027266
Silicon, disks, 13mm, thickness 0.38mm, single crystal, 100%
Si