EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Liquiritigenin inhibits tumor growth and vascularization in a mouse model of HeLa cells.
PMID 22692244
Abstract
Angiogenesis is one of the crucial steps in the transition of a tumor from a small, harmless cluster of mutated cells to a large, malignant growth, capable of spreading to other organs throughout the body. Vascular endothelial growth factor (VEGF) that stimulates vasculogenesis and angiogenesis is thought to be as an anti-angiogenic target for cancer therapy. Liquiritigenin (LQ), a flavanone existing in Radix glycyrrhiza, shows extensive biological activities, such as anti-inflammatory and anti-cancer properties. In our studies, liquiritigenin effectively inhibited the growth of tumors xenografted in nude mice from human cervical cancer cell line HeLa cells, and microvascular density (MVD) of the tumor exposed to liquiritigenin was reduced in a dose dependent manner, especially in the high dose group. Moreover, the expression and secretion of VEGF were down-regulated by the drug in vivo and in vitro. Therefore, liquiritigenin can be further studied on cancer and other diseases associated with VEGF up-regulation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
78825
Liquiritigenin, ≥97.0% (HPLC)
C15H12O4