EMAIL THIS PAGE TO A FRIEND
Nursing
New drugs 2012, part 2.
PMID 22692631
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H157
HOE 140, ≥94%
C59H89N19O13S