EMAIL THIS PAGE TO A FRIEND
Clinical orthopaedics and related research
Case report: High chromium and cobalt levels in a pregnant patient with bilateral metal-on-metal hip arthroplasties.
PMID 22692823
Abstract
Metal-on-metal bearings frequently are used in young patients leading to the concern that disseminated metals such as chromium (Cr) and cobalt (Co) as the main constituents could affect pregnancies. We describe a 41-year-old patient with bilateral metal-on-metal hip arthroplasties, a recurrent pseudotumor, and extremely high blood levels (Cr 39 μg/L, Co 138 μg/L) at 12 gestational weeks. At different gestational weeks, maternal blood, aspirate of the pseudotumor, and amniotic fluid were analyzed for Cr and Co. Therapy with chelating agents was not recommended because the mother showed no symptoms of toxicity and the safety of chelating therapy during pregnancy is not established. At 38 weeks of gestation, a healthy male infant was delivered with elevated Cr and Co cord blood levels. At the age of 8 weeks, the infant's Cr was comparable to the cord blood level, whereas the Co decreased considerably without treatment. At the age of 14 weeks, the infant's development was seemingly uneventful and no signs of toxicity were obvious. Carcinogenic, mutagenic, and teratogenic potentials of these metals have been suggested. However, we found no published clinical observations in context with pregnancies of women with hip arthroplasties using metal-on-metal implants. To our knowledge, this is the first report of such high levels of Cr and Co in a human pregnancy. Although we cannot generalize from one case, the seemingly uneventful outcome of this pregnancy may reassure colleagues when counseling patients with high ion levels whether to carry a pregnancy to term.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF81420094
Chromium, foil, 50x50mm, thickness 2.0mm, hard, 99.7+%
Cr
GF07622759
Chromium, foil, 50x50mm, thickness 3.0mm, hard, 99.95%
Cr
GF95051369
Chromium, foil, 25x25mm, thickness 2.0mm, hard, 99.7+%
Cr
GF15403047
Chromium, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 99.99+%
Cr
GF59666945
Chromium, rod, 10mm, diameter 5.0mm, 99.7+%
Cr
GF88955410
Chromium, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.99+%
Cr
GF15970080
Chromium, foil, light tested, 25x25mm, thickness 0.04mm, permanent polyester support, 99.99+%
Cr
GF50053092
Chromium, rod, 25mm, diameter 5.0mm, 99.7+%
Cr
GF22749240
Chromium, rod, 10mm, diameter 2.0mm, 99.7+%
Cr
GF84397168
Chromium, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%
Cr
GF97385338
Chromium, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.99+%
Cr
GF23770885
Chromium, foil, not light tested, 100x100mm, thickness 0.03mm, permanent polyester support, 99.99+%
Cr
GF24605816
Chromium, foil, not light tested, 100x100mm, thickness 0.04mm, permanent polyester support, 99.99+%
Cr
GF00270593
Chromium, foil, not light tested, 25x25mm, thickness 0.0015mm, permanent polyester support, 99.99+%
Cr
GF94865170
Chromium, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 99.99+%
Cr
GF73692377
Chromium, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 99.99+%
Cr
GF61658725
Chromium, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 99.99+%
Cr
GF81124286
Chromium, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 99.99+%
Cr
GF05241316
Chromium, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 99.99+%
Cr
GF98123250
Chromium, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 99.99+%
Cr
GF34438923
Chromium, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 99.99+%
Cr
GF49392456
Chromium, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 99.99+%
Cr
GF88985446
Chromium, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 99.99+%
Cr
GF30096053
Chromium, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Cr
GF12477157
Chromium, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.99+%
Cr
GF50354692
Chromium, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.99+%
Cr
GF74347050
Chromium, foil, not light tested, 25x25mm, thickness 0.03mm, permanent polyester support, 99.99+%
Cr
GF35390651
Chromium, foil, light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Cr
GF46122645
Chromium, foil, not light tested, 25x25mm, thickness 0.04mm, permanent polyester support, 99.99+%
Cr
GF31792956
Chromium, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.99+%
Cr