EMAIL THIS PAGE TO A FRIEND
Journal of Asian natural products research
Activation of ATM-Chk2 by 16-dehydropregnenolone induces G1 phase arrest and apoptosis in HeLa cells.
PMID 22694166
Abstract
The natural pregnane steroid 16-dehydropregnenolone (16-DHP) showed the growth inhibitory activity against human tumor cells; however, the mechanisms of actions of 16-DHP were not clarified. In this study, 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide assay was used to investigate the growth inhibitory effect of 16-DHP. Cell cycle distribution was analyzed using flow cytometry. Hoechst 33258 staining and DNA agarose gel electrophoresis were used to detect apoptosis. The levels of proteins were probed by Western blotting, and caspase activities were analyzed using Caspase Activity Kit. We found that 16-DHP inhibited the growth of human cervical carcinoma cells (HeLa cells) in a time- and dose-dependent manner. The growth inhibitory effect of 16-DHP was associated with G1 arrest mediated by ataxia telangiectasia mutated (ATM)-checkpoint kinase 2 (Chk2)-p53 signaling, as demonstrated by induction of the phosphorylations of ATM, Chk2, and p53 proteins. Followed by G1 arrest, 16-DHP-treated HeLa cells underwent caspase-dependent apoptosis. The inhibitors of caspase-3 and caspase-9 but not caspase-8 inhibitor blocked 16-DHP-induced apoptosis. Moreover, 16-DHP increased the level of Bax protein and the release of cytochrome c from mitochondria, but had no effect on the level of Bcl-2. These results suggested that 16-DHP inhibited the growth of HeLa cells via inducing ATM-Chk2-p53 activation-mediated G1 arrest and mitochondrial cell apoptosis.