EMAIL THIS PAGE TO A FRIEND
Chemosphere
Photocatalytic debromination of preloaded decabromodiphenyl ether on the TiO(2) surface in aqueous system.
PMID 22694774
Abstract
There have been serious concerns about polybromodiphenyl ethers (PBDEs) in the environment because of their global distribution and bioaccumulation. Owing to strong hydrophobicity of PBDEs, the regular photocatalytic system, in which the substrate is solvated in the bulk solution, is not applicable to the removal of the PBDEs in water. In this work, the photocatalytic reduction degradation of decabromodiphenyl ether (BDE209), the most-used PBDEs, was investigated in aqueous system, by pre-adsorbing it on the surface of photocatalyst. It was found that the preloaded BDE209 underwent efficient reductive debromination in aqueous system under irradiation with wavelength larger than 360 nm in the presence of electron donors such as methanol. Our experiments further show that such a preloaded system exhibits different characteristics from that in the organic solution. The meta-debrominated intermediate is predominant in the present system, while the ortho-debrominated one is the main nona-BDE products in the organic solution. In addition, different from other photocatalytic system, the pH has little effect on the photocatalytic reaction. We propose that these differences are originated from the formation of overlayer of hydrophobic BDE209 to limit the motion of BDE209 and the access of water and H(+)/OH(-) to the TiO(2) surface.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34120
BDE No 209 solution, 50 μg/mL in isooctane:toluene (9:1), analytical standard
C12Br10O