EMAIL THIS PAGE TO A FRIEND
Journal of fluorescence
Synthesis and characterization of 8-hydroxyquinoline complexes of tin(IV) and their application in organic light emitting diode.
PMID 22695927
Abstract
A series of 8-hydroxyquinoline complexes of tin, Q(2)SnCl(2) (Q = 2-methyl-8-hydroxyquinoline, 8-hydroxyquinoline, 5,7-dibromo-8-hydroxyquinoline, 5-chloro-8-hydroxyquinoline, 5,7-dichloro-8-hydroxyquinoline and 5-nitro-8-hydroxyquinoline) were prepared by reacting stannous dichloride with 8-hydroxyquinoline and its derivatives. All complexes were characterized by elemental analysis, mass spectrometry and infrared, UV-vis and (1)H NMR spectroscopes. Furthermore, the molecular structure of a representative complex, dichlorido-bis(5-nitro-quinolin-8-olato-2N,O)tin(IV), was determined by single-crystal X-ray diffraction. The photoluminescence (PL) properties of all prepared compounds and electroluminescence (EL) property of a selected complex (Q = 5-chloro-8-hydroxyquinoline) were investigated. The results showed that the emission wavelength can be tuned by electron donating or withdrawing group substituent on 8-hydroxyquinoline. Application of prepared complexes in fabrication of an OLED has been demonstrated.